missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 228340010

 View more versions of this product

Catalog No. AC228340010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Brij™ 35
5mg KOH/g max.
Waxy Pellets
33.0°C to 40.0°C
>149°C
Authentic
C14H30O2
C12H25(OCH2CH2)23OH
Polyoxyethylene(23)lauryl ether
SFNALCNOMXIBKG-UHFFFAOYSA-N
2-(dodecyloxy)ethan-1-ol
1199.56
White
9002-92-0
>100.0°C
1 kg
40 to 60mg KOH/g
HDPE plastic bottle
1% max.
MFCD00043063
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, propylene glycol, insoluble in mineral oil and vegetable oils
CCCCCCCCCCCCOCCO
230.39

Chemical Identifiers

9002-92-0
230.39
SFNALCNOMXIBKG-UHFFFAOYSA-N
2-(dodecyloxy)ethan-1-ol
C14H30O2
MFCD00043063
Polyoxyethylene(23)lauryl ether
CCCCCCCCCCCCOCCO
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only