missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 228340051
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Brij™ 35 | |
5mg KOH/g max. | |
Waxy Pellets | |
33.0°C to 40.0°C | |
>149°C | |
Authentic | |
C14H30O2 | |
C12H25(OCH2CH2)23OH | |
Polyoxyethylene(23)lauryl ether | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol | |
1199.56 |
White | |
9002-92-0 | |
>100.0°C | |
5 kg | |
40 to 60mg KOH/g | |
Plastic drum | |
1% max. | |
MFCD00043063 | |
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, propylene glycol, insoluble in mineral oil and vegetable oils | |
CCCCCCCCCCCCOCCO | |
230.39 |
Chemical Identifiers
9002-92-0 | |
230.39 | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol |
C14H30O2 | |
MFCD00043063 | |
Polyoxyethylene(23)lauryl ether | |
CCCCCCCCCCCCOCCO |
RUO – Research Use Only