missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brij∣r L23, Electrophoresis Reagent, Thermo Scientific Chemicals

$51.42 - $114.34

Specifications

Chemical Name or Material Brij™ L23
Color White
Name Note Electrophoresis Reagent
CAS 9002-92-0
Physical Form Waxy Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6609630
View Documents
Thermo Scientific Chemicals
J6609630
250 g
Each for $51.42
 
AAJ660960B
View Documents
Thermo Scientific Chemicals
J660960B
1000 g
Each for $114.34
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Brij™ L23
Electrophoresis Reagent
Waxy Solid
MFCD00080891
Soluble in water
White
9002-92-0
∼35°C to 40°C
Brij™ 35; Tricosaethylene glycol mono-n-dodecyl ether
1199.57
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only