Learn More
Brij∣r L23
CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
$65.42 - $539.58
Chemical Identifiers
| CAS | 9002-92-0 |
|---|---|
| Molecular Formula | C14H30O2 |
| Molecular Weight (g/mol) | 230.39 |
| MDL Number | MFCD00043063 |
| InChI Key | SFNALCNOMXIBKG-UHFFFAOYSA-N |
| Synonym | Brij™ 35; Tricosaethylene glycol mono-n-dodecyl ether |
| IUPAC Name | 2-(dodecyloxy)ethan-1-ol |
| SMILES | CCCCCCCCCCCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1580930
|
Thermo Scientific Chemicals
A1580930 |
250 g |
Each for $65.42
|
|
|||||
|
AAA158090B
|
Thermo Scientific Chemicals
A158090B |
1000 g |
Each for $133.66
|
|
|||||
|
AAA158090I
|
Thermo Scientific Chemicals
A158090I |
5000 g |
Each for $539.58
|
|
|||||
Description
Brij™ L23 is use in Stein-Moore chromatography. Used in the schlerotherapy treatments, in those suffering from blood vessel malformations and mutations. It acts as a surfactant in Micellar electrokinetic chromatography (MEKC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (40 mg/ml) at 20°C methanol, chloroform and ethanol.
Notes
Store at room temperature. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent. Used as a wetting agent, a detergent, or an emulsifier. Brij™L23 is a detergent used for measuring the concentration of carbon black in lung tissues.
Chemical Identifiers
| 9002-92-0 | |
| 230.39 | |
| SFNALCNOMXIBKG-UHFFFAOYSA-N | |
| 2-(dodecyloxy)ethan-1-ol |
| C14H30O2 | |
| MFCD00043063 | |
| Brij™ 35; Tricosaethylene glycol mono-n-dodecyl ether | |
| CCCCCCCCCCCCOCCO |
Specifications
| 9002-92-0 | |
| 250 g | |
| CH3(CH2)10CH2(OCH2CH2)23OH | |
| Soluble in water (40mg/ml) at 20°C methanol,chloroform and ethanol. | |
| CCCCCCCCCCCCOCCO | |
| 230.39 | |
| Brij 35 |
| ∽35°C to 40°C | |
| C14H30O2 | |
| MFCD00043063 | |
| SFNALCNOMXIBKG-UHFFFAOYSA-N | |
| 2-(dodecyloxy)ethan-1-ol | |
| 1199.57 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 500-002-6
RTECSNumber : JR5990000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only