missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Brilliant Stain Buffer DFS Item

For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension. Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample.

$226.00 - $1,064.00

Specifications

Chemical Name or Material Staining Buffer
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
00440942 DFS Item
View Documents
Invitrogen™
00440942
100 Tests N/A
 
00440975 DFS Item
View Documents
Invitrogen™
00440975
1000 Tests N/A
 
Description

Description

  • For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension
  • Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample
  • Prevents non-specific polymer interactions that can result in data appearing under-compensated
  • Buffer is provided in a 50 μL/test format
  • Compatible with traditional fluorochromes, Brilliant Violet dyes, Brilliant Ultra Violet dyes, Super Bright dyes, and standard flow cytometry protocols
  • Buffer is not compatible with UltraComp eBeads and UltraComp eBeads Plus compensation beads
  • If using these beads as a compensation tool, Flow Cytometry Staining Buffer should be used exclusively for all dilutions and washes during staining of the microspheres
Specifications

Specifications

Staining Buffer
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'