missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Brimonidine, 98%
Chemical Identifiers
CAS | 59803-98-4 |
---|---|
Molecular Formula | C11H10BrN5 |
Molecular Weight (g/mol) | 292.14 |
MDL Number | MFCD00153878 |
InChI Key | XYLJNLCSTIOKRM-UHFFFAOYSA-N |
Synonym | brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline |
PubChem CID | 2435 |
ChEBI | CHEBI:3175 |
IUPAC Name | 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine |
SMILES | BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC461522500
|
Thermo Scientific™
461522500 |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC461520010
|
Thermo Scientific™
461520010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
59803-98-4 | |
292.14 | |
XYLJNLCSTIOKRM-UHFFFAOYSA-N | |
2435 | |
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine |
C11H10BrN5 | |
MFCD00153878 | |
brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline | |
CHEBI:3175 | |
BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1 |
Specifications
59803-98-4 | |
Authentic | |
C11H10BrN5 | |
250mg | |
Solubility in water: insoluble. | |
BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1 | |
292.14 | |
CHEBI:3175 | |
98% |
0.3% max. | |
97.5% min. (HPLC) | |
MFCD00153878 | |
brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline | |
XYLJNLCSTIOKRM-UHFFFAOYSA-N | |
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine | |
2435 | |
292.13 | |
Brimonidine |
Safety and Handling
GHS H Statement Toxic if swallowed.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
RUO â Research Use Only