missing translation for 'onlineSavingsMsg'
Learn More

Bromide Standard, 1000 ppm Br-, Ricca Chemical

Supplier:  Ricca Chemical Company 11804

 View more versions of this product

Catalog No. 1180-4


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bromide Standard
7647-15-6,7732-18-5
Liquid
MFCD00003475
100°C
[Na+].[Br-]
102.89
253881
Natural Poly Bottle
1 mL = 1 mg Br
BrNa
Colorless
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide
120 mL
CHEBI:63004

Chemical Identifiers

7647-15-6
102.89
JHJLBTNAGRQEKS-UHFFFAOYSA-M
CHEBI:63004
[Na+].[Br-]
BrNa
MFCD00003475
253881
sodium bromide
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions