Learn More
Bromine, 99+%, extra pure
CAS: 7726-95-6 | Br | 79.91 g/mol
$354.07 - $354.07
Chemical Identifiers
| CAS | 7726-95-6 |
|---|---|
| Molecular Formula | Br |
| Molecular Weight (g/mol) | 79.91 |
| MDL Number | MFCD00010896 |
| InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-M |
| Synonym | bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish |
| PubChem CID | 24408 |
| ChEBI | CHEBI:29224 |
| SMILES | [Br-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC402845000
|
Thermo Scientific Chemicals
402845000 |
500 mL | Plastic Coated Glass Bottle |
Each for $354.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7726-95-6 | |
| 79.91 | |
| CPELXLSAUQHCOX-UHFFFAOYSA-M | |
| 24408 | |
| [Br-] |
| Br | |
| MFCD00010896 | |
| bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
| CHEBI:29224 |
Specifications
| 58.7°C | |
| 7726-95-6 | |
| Fuming Liquid | |
| 99% min. (Iodometry) | |
| Br2 | |
| 03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47 | |
| bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
| CPELXLSAUQHCOX-UHFFFAOYSA-M | |
| bromide | |
| 24408 | |
| 159.82 | |
| Extra Pure | |
| 3.111 | |
| Bromine |
| -7.2°C | |
| Red to Brown | |
| 500 mL | |
| Br | |
| MFCD00010896 | |
| 15, 1401 | |
| Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2 | |
| [Br-] | |
| 79.91 | |
| CHEBI:29224 | |
| ≥99% | |
| Plastic Coated Glass Bottle | |
| 3.1110g/mL |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 231-778-1
RUO – Research Use Only