missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromine liquid, 99.8%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 00090530
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bromine, liquid | |
Liquid | |
99.8% | |
MFCD00010896 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
bromide | |
24408 | |
159.81 | |
3.1g/cm3 |
7726-95-6 | |
250 g | |
Br | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
[Br-] | |
79.91 | |
CHEBI:29224 | |
99.8% |
Chemical Identifiers
7726-95-6 | |
79.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
24408 | |
[Br-] |
Br | |
MFCD00010896 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CHEBI:29224 |
Safety and Handling
P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H330
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 231-778-1
RTECSNumber : EF9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only