missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromine liquid, ACS, 99.5%, Thermo Scientific Chemicals

Maximum level of impurities: Evaporation residue 0.005%, Cl 0.05%, I 0.001%, Sulfur compounds (as S) 0.001%, Heavy Metals (as Pb) 2ppm, Ni 5ppm

Supplier:  Thermo Scientific Chemicals 03322022

 View more versions of this product

Catalog No. AA3322022


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bromine liquid
Brown to Red
100 g
Br
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
[Br-]
79.91
CHEBI:29224
99.5%
Unpleasant
230 hPa (173mm Hg) at 20°C
7726-95-6
Liquid
99.5%
MFCD00010896
CPELXLSAUQHCOX-UHFFFAOYSA-M
bromide
24408
159.81
ACS
Red-brown fuming liquid
3.12g/cm3 at 20°C

Chemical Identifiers

7726-95-6
79.91
CPELXLSAUQHCOX-UHFFFAOYSA-M
24408
[Br-]
Br
MFCD00010896
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
CHEBI:29224
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H330

DOTInformation : DOT Class: 8; Packing Group: I

EINECSNumber : 231-778-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only