Learn More
Bromoacetaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
$73.89 - $838.04
Chemical Identifiers
CAS | 2032-35-1 |
---|---|
Molecular Formula | C6H13BrO2 |
Molecular Weight (g/mol) | 197.072 |
MDL Number | MFCD00000214 |
InChI Key | LILXDMFJXYAKMK-UHFFFAOYSA-N |
Synonym | bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal |
PubChem CID | 74852 |
IUPAC Name | 2-bromo-1,1-diethoxyethane |
SMILES | CCOC(CBr)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1327622
|
Thermo Scientific Chemicals
A1327622 |
100 g |
Each for $73.89
|
|
|||||
AAA1327636
|
Thermo Scientific Chemicals
A1327636 |
500 g |
Each for $231.05
|
|
|||||
AAA132760E
|
Thermo Scientific Chemicals
A132760E |
2500 g |
Each for $838.04
|
|
|||||
Description
Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ?-heterosubstituted acetaldehyde acetals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other É-heterosubstituted acetaldehyde acetals.
Solubility
It is immiscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Protect from frost, heat and sunlight. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Acids. Moisture Sensitive.
Chemical Identifiers
2032-35-1 | |
197.072 | |
LILXDMFJXYAKMK-UHFFFAOYSA-N | |
74852 | |
CCOC(CBr)OCC |
C6H13BrO2 | |
MFCD00000214 | |
bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal | |
2-bromo-1,1-diethoxyethane |
Specifications
2032-35-1 | |
66°C to 67°C (18 mmHg) | |
C6H13BrO2 | |
BrCH2CH(OCH2CH3)2 | |
100 g | |
635754 | |
bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal | |
LILXDMFJXYAKMK-UHFFFAOYSA-N | |
2-bromo-1,1-diethoxyethane | |
74852 | |
97% |
1.27 | |
61°C (142°F) | |
1.439 | |
MFCD00000214 | |
UN2810 | |
Moisture sensitive | |
It is immiscible with water. | |
CCOC(CBr)OCC | |
197.072 | |
197.08 | |
Bromoacetaldehyde diethyl acetal |
Safety and Handling
GHS H Statement
H301-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
H227-H301-H315-H319-H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 217-989-1
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only