Learn More
Bromoacetyl bromide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1958014
Description
Bromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.
Solubility
Immiscible with water.
Notes
Moisture Sensitive. Store in cool place. Hydrolyses readily. Incompatible with strong oxidizing agents, alcohols and strong bases.
Specifications
Bromoacetyl bromide | |
2.324 | |
>105°C (221°F) | |
C2H2Br2O | |
MFCD00000115 | |
UN2513 | |
Moisture sensitive | |
Immiscible with water. | |
C(C(=O)Br)Br | |
201.845 | |
201.86 |
598-21-0 | |
147°C to 150°C | |
Acrid | |
1.546 | |
25 g | |
605440 | |
bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide | |
LSTRKXWIZZZYAS-UHFFFAOYSA-N | |
2-bromoacetyl bromide | |
61147 | |
98% |
Chemical Identifiers
598-21-0 | |
201.845 | |
LSTRKXWIZZZYAS-UHFFFAOYSA-N | |
61147 | |
C(C(=O)Br)Br |
C2H2Br2O | |
MFCD00000115 | |
bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide | |
2-bromoacetyl bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BROMOACETYL BROMIDE
EINECSNumber : 209-923-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only