Learn More
Thermo Scientific Chemicals Bromocresol Green, pure, indicator grade
Bromocresol Green, C21H14Br4O5S, CAS Number-76-60-8, 3', 3″, 5', 3',3″,5',5″-Tetrabromo-m-cresolsulfonephthalein, BCG, 5g, Brown, 200-972-8, 698.01, Pure, Warning, GHS H Statement: May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
$98.40 - $244.42
Chemical Identifiers
CAS | 76-60-8 |
---|---|
Molecular Formula | C21H14Br4O5S |
Molecular Weight (g/mol) | 698.014 |
MDL Number | MFCD00005874 |
InChI Key | FRPHFZCDPYBUAU-UHFFFAOYSA-N |
Synonym | 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' |
PubChem CID | 6451 |
IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC151310050
|
Thermo Scientific Chemicals
151310050 |
5 g | Glass bottle |
Each for $98.40
|
|
||||
AC151310250
|
Thermo Scientific Chemicals
151310250 |
25 g | Glass bottle |
Each for $244.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
76-60-8 | |
698.014 | |
FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
6451 | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
C21H14Br4O5S | |
MFCD00005874 | |
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' | |
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
Specifications
225°C, 437°F | |
C21H14Br4O5S | |
15, 1393 | |
Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethyl acetate | |
Authentic | |
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol | |
6451 | |
Pure | |
600 min. (617nm, in buffer) | |
Brown | |
Powder |
76-60-8 | |
MFCD00005874 | |
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5' | |
FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br | |
698.014 | |
698.01 | |
Glass bottle | |
from pH 3.8 (yellow) to pH 5.4 (blue) | |
5 g | |
Bromocresol Green |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 200-972-8
RTECSNumber : SJ7456000
TSCA : TSCA
RUO – Research Use Only