missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromocresol Green, Reagent, ACS, Spectrum™ Chemical
BR132, 76-60-8, 62625-32-5, C21H14Br4O5S
Supplier: Spectrum Chemical Mfg Cor BR1321KGBL
Description
Spectrum™ Chemical Bromocresol Green, Reagent, ACS , is a dye in the triphenylmethane family and is used as a tracking dye and pH indicator for DNA agarose gel electrophoresis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
76-60-8 | |
C21H14Br4O5S | |
CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C | |
698.01 | |
Amber Glass Bottle |
100% | |
FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione | |
ACS | |
1 kg |
Chemical Identifiers
76-60-8 | |
698.01 | |
3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
C21H14Br4O5S | |
FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C |