missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green TS, 0.05% (w/v) Alcoholic Solution, Ricca Chemical
Supplier: Ricca Chemical Company 12024
Specifications
| Bromocresol Green | |
| 67-17-5,67-56-1,76-60-8 | |
| 100.0,0.06 | |
| Indicator | |
| Alcoholic Solution | |
| 120 mL |
| Test Solution | |
| 98.0,0.06 | |
| 0.05% (w/v) | |
| Natural Poly Bottle | |
| Orange | |
| Liquid |