missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Bromocresol Purple, pure, indicator grade

Supplier:  Thermo Scientific Chemicals 151330050

 View more versions of this product

Catalog No. AC151330050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

(20 mg/L in methanol) Complies, (20 mg/L in methanol) Complies at 0nm
245.0°C
C21H16Br2O5S
Bromcresol Purple, BCP, 5', 5''-Dibromo-o-cresolsulfonephthalein
ABIUHPWEYMSGSR-UHFFFAOYSA-N
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
540.22
CHEBI:86154
Pure
from pH 5.2 (yellow) to pH 6.8 (purple)
5 g
Bromocresol Purple
115-40-2
MFCD00011681
Solubility in water: practically insoluble. Other solubilities: 75g/L in ethanol (20°C)
Authentic
3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
8273
540.21
Glass bottle
Pink or Salmon
Powder

Chemical Identifiers

115-40-2
540.22
ABIUHPWEYMSGSR-UHFFFAOYSA-N
8273
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
C21H16Br2O5S
MFCD00011681
Bromcresol Purple, BCP, 5', 5''-Dibromo-o-cresolsulfonephthalein
CHEBI:86154
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only