missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromocresol Purple TS, 0.1% (w/v) in dilute Sodium Hydroxide, Ricca Chemical
Supplier: Ricca Chemical Company 131016
Specifications
Bromocresol Purple | |
98.0,0.09,0.03 | |
115-40-2,1310-73-2 | |
Natural Poly Bottle | |
Purple | |
Liquid |
Test Solution | |
100.0,0.1,0.03 | |
0.1% (w/v) | |
Dilute Sodium Hydroxide | |
500 mL |