missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromocresol Purple TS, 0.1% (w/v) in dilute Sodium Hydroxide, Ricca Chemical

Supplier:  Ricca Chemical Company RI13104

 View more versions of this product

Catalog No. 1310-4


Explore available promotions

Description

Description

Specifications

Specifications

Bromocresol Purple
98.0,0.09,0.03
115-40-2,1310-73-2
Natural Poly Bottle
Purple
Liquid
Test Solution
100.0,0.1,0.03
0.1% (w/v)
Dilute Sodium Hydroxide
120 mL
Videos
SDS
Documents

Documents

Promotions

Promotions