missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromocyclopentane, 98+%, Thermo Scientific Chemicals

$97.16 - $321.41

Chemical Identifiers

CAS 137-43-9
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00001359
InChI Key BRTFVKHPEHKBQF-UHFFFAOYSA-N
Synonym cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
PubChem CID 8728
IUPAC Name bromocyclopentane
SMILES C1CCC(C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1545522
View Documents
Thermo Scientific Chemicals
A1545522
100 g
Each for $97.16
 
AAA1545536
View Documents
Thermo Scientific Chemicals
A1545536
500 g
Each for $321.41
 
Description

Description

Bromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

137-43-9
149.031
BRTFVKHPEHKBQF-UHFFFAOYSA-N
8728
C1CCC(C1)Br
C5H9Br
MFCD00001359
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
bromocyclopentane

Specifications

137-43-9
137°C to 139°C
98%
1.4895
100 g
1209256
Immiscible with water.
C1CCC(C1)Br
149.031
149.04
Liquid
1.390
42°C (107°F)
C5H9Br
MFCD00001359
UN1993
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
BRTFVKHPEHKBQF-UHFFFAOYSA-N
bromocyclopentane
8728
≥98%
Bromocyclopentane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 205-294-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only