missing translation for 'onlineSavingsMsg'
Learn More

(Bromodifluoromethyl)trimethylsilane 98.0+%, TCI America™

$164.03 - $497.70

Chemical Identifiers

CAS 115262-01-6
Molecular Formula C4H9BrF2Si
Molecular Weight (g/mol) 203.102
MDL Number MFCD18641931
InChI Key WDZVWBWAUSUTTO-UHFFFAOYSA-N
Synonym Bromodifluoro(trimethylsilyl)methane
PubChem CID 14639950
IUPAC Name [bromo(difluoro)methyl]-trimethylsilane
SMILES C[Si](C)(C)C(F)(F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B43251G
View Documents
TCI America
B43251G
1 g
Each for $164.03
Only null left
 
B43255G
View Documents
TCI America
B43255G
5 g
Each for $497.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

115262-01-6
203.102
WDZVWBWAUSUTTO-UHFFFAOYSA-N
14639950
C[Si](C)(C)C(F)(F)Br
C4H9BrF2Si
MFCD18641931
Bromodifluoro(trimethylsilyl)methane
[bromo(difluoro)methyl]-trimethylsilane

Specifications

115262-01-6
108°C
MFCD18641931
1993
WDZVWBWAUSUTTO-UHFFFAOYSA-N
[bromo(difluoro)methyl]-trimethylsilane
14639950
≥98.0% (GC)
(Bromodifluoromethyl)trimethylsilane
Colorless
C4H9BrF2Si
1 g
Bromodifluoro(trimethylsilyl)methane
C[Si](C)(C)C(F)(F)Br
203.102
203.10
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'