missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Bromomethyl)cyclobutane, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 367641000

 View more versions of this product

Catalog No. AC367641000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

(Bromomethyl)cyclobutane
17247-58-4
100.0
145.0°C to 148.0°C
Authentic
Glass bottle
1.4790 to 1.4830
MFCD00040960
05, 22
bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan
FLHFTXCMKFVKRP-UHFFFAOYSA-N
bromomethylcyclobutane
2734658
99%
99%
98.5
1.3390g/mL
27°C
98.5% min. (GC)
C5H9Br
C4H7CH2Br
100mL
1.339
Solubility in water: .. Other solubilities: soluble in alcohols
C1CC(C1)CBr
149.03
149.03

Chemical Identifiers

17247-58-4
149.03
FLHFTXCMKFVKRP-UHFFFAOYSA-N
2734658
C1CC(C1)CBr
C5H9Br
MFCD00040960
bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan
bromomethylcyclobutane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only