Learn More
Bromomethyl methyl ether, tech. 90%
CAS: 13057-17-5 | C2H5BrO | 124.965 g/mol
Supplier: Thermo Scientific Chemicals L0983714
Description
Bromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.
Solubility
Sparingly soluble in water.
Notes
Moisture Sensitive. Store at 4°C. Do not store together with acids. Store away from water and strong bases. Incompatible with acids, bases, moisture, heat and oxidizing agents.
Specifications
| Bromomethyl methyl ether | |
| 1.531 | |
| 26°C (78°F) | |
| C2H5BrO | |
| MFCD00000171 | |
| UN1992 | |
| Moisture sensitive | |
| Sparingly soluble in water. | |
| COCBr | |
| 124.965 | |
| 124.97 | |
| Technical |
| 13057-17-5 | |
| 86°C to 87°C | |
| Pungent | |
| 1.4520-1.4600 | |
| 25 g | |
| 1730853 | |
| bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether | |
| JAMFGQBENKSWOF-UHFFFAOYSA-N | |
| bromo(methoxy)methane | |
| 83093 | |
| 90% |
Chemical Identifiers
| 13057-17-5 | |
| 124.965 | |
| JAMFGQBENKSWOF-UHFFFAOYSA-N | |
| 83093 | |
| COCBr |
| C2H5BrO | |
| MFCD00000171 | |
| bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether | |
| bromo(methoxy)methane |
Safety and Handling
GHS H Statement
H226-H351-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P363-P370+P378q-P5
H226-H302+H312+H332-H315-H319-H335-H351
DOTInformation : Hazard Class: 3; Packaging Group: III
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only