Learn More
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
$192.73 - $192.73
Chemical Identifiers
| CAS | 563-70-2 |
|---|---|
| Molecular Formula | CH2BrNO2 |
| Molecular Weight (g/mol) | 139.94 |
| MDL Number | MFCD00007401 |
| InChI Key | DNPRVXJGNANVCZ-UHFFFAOYSA-N |
| Synonym | methane, bromonitro, bromo nitro methane, nitrobromomethane, unii-eg63p9eur0, eg63p9eur0, ccris 9456, bromo-nitromethane, 1-bromonitromethane, methane,bromonitro, monobromonitromethane |
| PubChem CID | 68423 |
| IUPAC Name | bromo(nitro)methane |
| SMILES | C([N+](=O)[O-])Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC307050100
|
Thermo Scientific Chemicals
307050100 |
10 g | Glass bottle |
Each for $192.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 563-70-2 | |
| 139.94 | |
| DNPRVXJGNANVCZ-UHFFFAOYSA-N | |
| 68423 | |
| C([N+](=O)[O-])Br |
| CH2BrNO2 | |
| MFCD00007401 | |
| methane, bromonitro, bromo nitro methane, nitrobromomethane, unii-eg63p9eur0, eg63p9eur0, ccris 9456, bromo-nitromethane, 1-bromonitromethane, methane,bromonitro, monobromonitromethane | |
| bromo(nitro)methane |
Specifications
| 563-70-2 | |
| 2.0700g/mL | |
| 113°C | |
| 88% min. (GC) | |
| CH2BrNO2 | |
| BrCH2NO2 | |
| 10 g | |
| methane, bromonitro, bromo nitro methane, nitrobromomethane, unii-eg63p9eur0, eg63p9eur0, ccris 9456, bromo-nitromethane, 1-bromonitromethane, methane,bromonitro, monobromonitromethane | |
| C([N+](=O)[O-])Br | |
| 139.94 | |
| 139.94 | |
| Technical | |
| Bromonitromethane, tech., 90% |
| Yellow | |
| 146°C to 148°C (750.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4850 to 1.495 | |
| MFCD00007401 | |
| 2.07 | |
| DNPRVXJGNANVCZ-UHFFFAOYSA-N | |
| bromo(nitro)methane | |
| 68423 | |
| 90% | |
| Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the ris
GHS Signal Word: Danger
EINECSNumber : 209-258-0
RTECSNumber : PA5360000
TSCA : TSCA
RUO – Research Use Only