missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromophenol Blue Solution, 0.04%/Certified, Fisher Chemical
Supplier: Fisher Chemical SI12500
Description
Solution of bromophenol blue sodium salt for colorimetric determinations.EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Bromophenol Blue Solution, 0.04% | |
0.04% | |
MFCD00013793 | |
0.04% | |
Pass test | |
500 mL |
62625-28-9,7732-18-5 | |
0.04% | |
Certified | |
Glass Bottle | |
Blue | |
Liquid |