missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromoterephthalic Acid 98.0+%, TCI America™
$326.98 - $1186.84
Chemical Identifiers
CAS | 586-35-6 |
---|---|
Molecular Formula | C8H5BrO4 |
Molecular Weight (g/mol) | 245.028 |
MDL Number | MFCD00002403 |
InChI Key | QPBGNSFASPVGTP-UHFFFAOYSA-N |
Synonym | bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9 |
PubChem CID | 68513 |
IUPAC Name | 2-bromoterephthalic acid |
SMILES | C1=CC(=C(C=C1C(=O)O)Br)C(=O)O |
Chemical Identifiers
586-35-6 | |
245.028 | |
QPBGNSFASPVGTP-UHFFFAOYSA-N | |
68513 | |
C1=CC(=C(C=C1C(=O)O)Br)C(=O)O |
C8H5BrO4 | |
MFCD00002403 | |
bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9 | |
2-bromoterephthalic acid |
Specifications
586-35-6 | |
C8H5BrO4 | |
5 g | |
QPBGNSFASPVGTP-UHFFFAOYSA-N | |
2-bromoterephthalic acid | |
68513 | |
≥98.0% (GC) | |
Bromoterephthalic Acid |
White-Yellow | |
MFCD00002403 | |
bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9 | |
C1=CC(=C(C=C1C(=O)O)Br)C(=O)O | |
245.028 | |
245.03 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0062
TSCA : No