missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Indicator, 0.04% (w/v), Ricca Chemical

For Glassware Residue Detection

Supplier:  Ricca Chemical Company 137516

 View more versions of this product

Catalog No. 1375-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bromothymol Blue Indicator
98.0,0.04
0.04% (w/v)
Indicator
Aqueous Solution
6.70 to 6.90
Liquid
34722-90-2,7732-18-5
100.0,0.04
0.04% W/V
Natural Poly Bottle
Dark Blue
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions