missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromothymol Blue Solution (0.04%/Certified), Fisher Chemical
Supplier: Fisher Chemical SI22500
Description
Solution of bromothymol blue sodium salt for colorimetric determinations.EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Bromothymol Blue Solution, 0.04% | |
34722-90-2,7732-18-5,64-02-8 | |
0.04% | |
Certified | |
Glass Bottle | |
Pass test | |
Green | |
Liquid |
0°C | |
0.04% | |
MFCD00077263 | |
0.04% | |
14mmHg at 20°C | |
1g/cm³ | |
500 mL |