Learn More
Brucine, 99%, anhydrous, Thermo Scientific Chemicals
$50.81 - $2241.64
Chemical Identifiers
CAS | 357-57-3 |
---|---|
Molecular Formula | C23H26N2O4 |
Molecular Weight (g/mol) | 394.46 |
MDL Number | MFCD00005942 |
InChI Key | RRKTZKIUPZVBMF-VLTSECPOSA-N |
Synonym | brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy |
PubChem CID | 51413923 |
IUPAC Name | (4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one |
SMILES | COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC278720050
|
Thermo Scientific Chemicals
278720050 |
5 g | Glass bottle |
Each for $50.81
|
|
||||
AC278720500
|
Thermo Scientific Chemicals
278720500 |
50 g | Glass bottle |
Each for $236.13
|
|
||||
AC278721000
|
Thermo Scientific Chemicals
278721000 |
100 g | Glass bottle |
Each for $366.66
|
|
||||
AC278725000
|
Thermo Scientific Chemicals
278725000 |
500 g | Glass bottle |
Each for $2,241.64
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
357-57-3 | |
394.46 | |
RRKTZKIUPZVBMF-VLTSECPOSA-N | |
51413923 | |
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC |
C23H26N2O4 | |
MFCD00005942 | |
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy | |
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one |
Specifications
357-57-3 | |
White to Beige | |
Authentic | |
Glass bottle | |
MFCD00005942 | |
27, IV, 7876 | |
−123° (20°C c=2.5,CCl3) | |
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy | |
RRKTZKIUPZVBMF-VLTSECPOSA-N | |
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one | |
51413923 | |
99% | |
Brucine, Anhydrous |
174.0°C to 179.0°C | |
1% max. (105°C, 3 hrs) | |
99% | |
C23H26N2O4 | |
5 g | |
15, 1464 | |
− 123.00 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, chloroform and benzene, slightly soluble in ether, glycerol and, ethyl acetate | |
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC | |
394.46 | |
394.46 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
Harmful to aquatic life with long lasting effects.
Fatal if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 206-614-7
RUO – Research Use Only