missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brucine dihydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1625118

 View more versions of this product

Catalog No. AAA1625118


Explore available promotions

Description

Description

Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

Solubility
Soluble in methanol. Partially soluble Miscible with water.

Notes
Incompatible with strong oxidizing agents, bases and strong reducing agents.
Specifications

Specifications

Brucine dihydrate
176°C to 178°C
MFCD00149384
UN1570
Soluble in methanol. Partially soluble Miscible with water.
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC.O.O
466.53
430.50 (394.47 Anhydrous)
5892-11-5
C23H34N2O8
50 g
brucine dihydrate, 10,11-dimethoxystrychnine, 10,11-dimethoxystrychnine dihydrate
XLXHHXCMBNBDMP-BEYGIVOKSA-N
(4aR,5aS,8aR,13aS,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;dihydrate
69897601
98%

Chemical Identifiers

5892-11-5
MFCD00149384
brucine dihydrate, 10,11-dimethoxystrychnine, 10,11-dimethoxystrychnine dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c

H300+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BRUCINE

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only