Learn More
Brucine sulfate heptahydrate, ACS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03624914
Description
As Resolving agent. Used in the Determination of nitrate. Used in Pharmaceutical and food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Brucine sulfate heptahydrate | |
180°C (decomposition) | |
MFCD00150159 | |
UN2811 | |
brucinesulfateheptahydrate fornitrateanalysis | |
PPJIVFYMRNHHTJ-UMRIXEKQSA-N | |
(4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate | |
126969999 | |
ACS |
60583-39-3 | |
C46H68N4O19S | |
25 g | |
14,1455 | |
Soluble in hot water. | |
O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34 | |
1013.12 | |
1013.15 (887.03 Anhydrous) | |
Powder |
Chemical Identifiers
60583-39-3 | |
1013.12 | |
PPJIVFYMRNHHTJ-UMRIXEKQSA-N | |
126969999 | |
O.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34 |
C46H68N4O19S | |
MFCD00150159 | |
brucinesulfateheptahydrate fornitrateanalysis | |
(4aR,5aR,8aR,13aR,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;heptahydrate |
Safety and Handling
GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
P260-P264b-P270-P271-P280-P301+P310-P304+P340-P310-P330-P501c
H300+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 225-432-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only