missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brucine sulfate hydrate, 98% (dry wt.), water <13%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1994322

 View more versions of this product

Catalog No. AAA1994322


Explore available promotions

Description

Description

Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Specifications

Brucine sulfate hydrate
191°C to 196°C (decomposition)
C46H54N4O12S
100 g
Hygroscopic
brucine sulfate salt
HCMSIGALSOEZRW-MDQLRNFDSA-N
10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;hydrate
118797651
98%
652154-10-4
(dry wt.), water <13%
MFCD00013472
UN2811
14,1455
Soluble in water.
OS(O)(=O)=O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1
887.01
443.52 (Anhydrous)

Chemical Identifiers

652154-10-4
887.01
HCMSIGALSOEZRW-MDQLRNFDSA-N
118797651
OS(O)(=O)=O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1
C46H54N4O12S
MFCD00013472
brucine sulfate salt
10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c

H300+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 225-432-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only