missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ BS3 (bis(sulfosuccinimidyl)suberate)
Crosslink amines to amines with BS3; composed of sulfo-NHS-ester groups separated by an 11.4A, water-soluble spacer arm that is useful for cell-surface protein crosslinking.
$339.09 - $3855.57
Specifications
Cleavable | No |
---|---|
Molecular Weight (g/mol) | 572.43 |
PEGylated | No |
Solubility | Water |
Product Type | Crosslinker |
Description
Thermo Scientific™ Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.
Highlights:
- Crosslink cell-surface proteins before cell lysis and immunoprecipitation
- Identify receptor-ligand interactions by chemical crosslinking
- “Fix” protein interactions to allow identification of weak or transient protein interactions
- Identify near-neighbor protein interactions
- Protein crosslinking to create bioconjugates via single-step reactions
- Immobilize proteins onto amine-coated surfaces
Specifications
No | |
No | |
Crosslinker |
572.43 | |
Water |
For Research Use Only. Not for use in diagnostic procedures.