missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

$507.68 - $1583.59

Specifications

Storage 2°C to 8°C
View More Specs

Products 5
Catalog Number Mfr. No. Standard Components Quantity Price Quantity  
Catalog Number Mfr. No. Standard Components Quantity Price Quantity  
PI38831
View Documents
Thermo Scientific™
TS38831
BSTFA +1% TMCS 10 x 1 mL ampules
Pack of 10 Each for $683.68
 
PI38832
View Documents
Thermo Scientific™
TS38832
BSTFA +1% TMCS 10 g
Pack for $507.68
 
PI38833
View Documents
Thermo Scientific™
TS38833
BSTFA +1% TMCS 25 g
Pack for $648.44
 
PI38834
View Documents
Thermo Scientific™
TS38834
BSTFA +1% TMCS 100 g
Each for $1,583.59
 
PI38840
View Documents
Thermo Scientific™
TS38840
BSTFA +10% TMCS 10 x 1 mL Ampules
Pack of 10 Each for $1,287.03
 
Description

Description

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone

Specifications

Specifications

2°C to 8°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.