missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ BSTFA Silylation Reagent

Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.

$655.44 - $1702.68

Specifications

Standard Concentration 98 to 100%
For Use With GC
Storage 2°C to 8°C
Standard Components Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI38828
View Documents
Thermo Scientific™
TS38828
25 g
Pack for $655.44
 
PI38829
View Documents
Thermo Scientific™
TS38829
100 g
Pack for $1,702.68
 
PI38830
View Documents
Thermo Scientific™
TS38830
10 x 1 mL ampules
Pack of 10 Each for $746.91
 
Description

Description

Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations. This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.

BSTFA is a powerful thrimethylsilyl donor, with donor strength that is comparable to its unfluorinated analog BSA [N,O-Bis(trimethylsilyl)acetamide]. BSTFA reacts to replace labile hydrogens on a wide range of polar compounds with a -Si(CH3)3 group. This physical characteristic is particularly useful in the gas chromatography of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
  • Increased volatility of reaction byproducts mono(trimethylsilyl)trifluoroacetamide and trifluoroacetamide over corresponding nonfluorinated compounds from BSA
  • Increased volatility makes it possible to derivatize smaller molecules with which the TMS derivatives elute with the byproducts from BSA

Specifications

Specifications

98 to 100%
GC
2°C to 8°C
Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.