BUCHI Boiling Point Tubes and Capillaries
Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.
Supplier:  BUCHI 051850
 View more versions of this product
Catalog No. 10-987-108
Explore available promotions
Description
Description
  • High precision for reliable resutls
  • Meets Siwoloboff and Eu Pharmacopeia standards
  • Designed for BUCHI melting and boiling point instruments
  • Compatible with other instruments
Specifications
Specifications
Boiling Point Capillaries
Borosilicate Glass 3.3
Autoclavable
0.8 mm
10/Pk.