Learn More
BUCHI Rotavapor Fill Tube
Transparent PTFE Tubing. Use to introduce solvent into evaporating flask during distillation.
Supplier:  BUCHI 000646
Catalog No. 05-422-892
Only null left
Explore available promotions
Description
Description
  • Transparent tubing PTFE
  • Ø4.7/5.5mm
  • Length 330mm
Specifications
Specifications
Transparent
PTFE
Introduction of solvent into evaporating flask during distillation
Tubing
Tubing
330 mm
Product Suggestions
Product Suggestions