missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agilent BioTek Buffer Bottle Assembly for 50 TS Microplate Washer

Accessory for 50 TS microplate washer

Supplier:  Agilent BioTek 4070515

Catalog No. BT4070515


Explore available promotions

Description

Description

  • For use with 50 TS Microplate Strip Washer
  • Includes: Bottle, cap w/fitting and tubing

Specifications

Specifications

2 L
67.62 oz.
ELx 405 Microplate Washer, Agilent BioTek Dispenser, Pipetting Systems with Non-autoclavable Fluid Paths
Buffer Bottle Assembly
Bottle, connector, cap with fitting and tubing
Videos
SDS
Documents

Documents

Promotions

Promotions