missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Buffer, pH 7.2, for Biochemical Oxygen Demand (BOD), Ricca Chemical
For Biochemical Oxygen Demand (BOD) Testing
$106.73 - $319.73
Specifications
| Color | Clear colorless |
|---|---|
| pH Range | 7.2 |
| Packaging | Natural Poly Bottle |
| Type | Phosphate Buffer |
Specifications
| Clear colorless | |
| 7.2 | |
| Natural Poly Bottle | |
| Phosphate Buffer |