missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 5.00 (Certified), Fisher Chemical™
Supplier: Fisher Chemical BPSB102500
Specifications
Buffer Solution, pH 5.00 | |
877-24-7,1310-73-2,7732-18-5 | |
5.0 | |
500 mL | |
MFCD00146206 | |
Traceable to NIST. |
0°C | |
100°C | |
Liquid | |
Pass Test | |
Poly Bottle | |
Certified |