missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 7.00 (Certified), Fisher Chemical™
Potassium Phosphate Monobasic/Sodium Hydroxide
Supplier: Thermo Fisher Scientific BPSB108500
Specifications
Buffer Solution, pH 7.00 | |
7778-77-0,1310-73-2,7732-18-5 | |
1.003g/cm³ | |
Liquid | |
Pass Test | |
Poly Bottle | |
14mmHg at 20°C | |
Certified |
0°C | |
Colorless | |
7.00 | |
500 mL | |
MFCD00134130 | |
Traceable to NIST. | |
Chlorine Titrimeter Reagent |