missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BugBuster™ Protein Extraction Reagent

Reagent for protein sample preparation

$154.00 - $1010.00

Specifications

Content And Storage Store at 15°C to 30°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
705844
View Documents
MilliporeSigma™
MIL705844
500 mL
Each for $598.00
Only null left
 
705843
View Documents
MilliporeSigma™
705843
100 mL
Each for $154.00
Only null left
 
70-584-1000ML
View Documents
MilliporeSigma™
705841000ML
1000 mL
Each for $1,010.00
Only null left
 
Description

Description

  • Formulated for gentle disruption of cell wall of E. coli, resulting in liberation of soluble protein
  • Provides simple, rapid, low cost alternative to mechanical methods
  • Formulation utilizes mixture of non-ionic detergents capable of cell wall perforation without denaturing soluble protein
Specifications

Specifications

Store at 15°C to 30°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'