missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications
Halogen Bulb | |
Halogen | |
12 V | |
Clear | |
1.27 cm | |
Microscope, surgical lights | |
1.17 in. | |
2.97 cm | |
T4 |
2 Pin (GY6.35) | |
2000 hr. | |
100 W | |
0.5 in. | |
CBAR6/C6F/Single Coil Flat/Oval Transverse | |
1.72 in. | |
4.3 cm | |
2800 lm |