Bulbtronics™ Halogen Light Bulbs DFS Item
Supplier:  Bulbtronics™ 0032368
 View more versions of this product
Catalog No. 14-395-892
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Description
Specifications
Specifications
Halogen Bulb
Halogen
12 V
1.12 in.
1.16 in.
800 lm
PY16-1.25
1000 hr.
35 W
2.85 cm
2.94 cm
T9