missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bulbtronics™ Osram Sylvania™ Xenon Arc Lamp
$709.44 - $1005.44
Catalog Number | Mfr. No. | Amperage | Base Style | Diameter (English) | Length (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Amperage | Base Style | Diameter (English) | Length (English) | Price | Quantity | ||
14395361
|
Bulbtronics™
251 |
25 A | SFA20-8, SFA20-10 | 1.14 in. | 10.24 in. |
Each for $709.44
|
|
||
14395593
|
Bulbtronics™
2628 |
5.4 A | SFA7.5-2, SFA7.5-2 | 0.39 in. | 3.54 in. |
Each for $1,005.44
|
|
||
14395592
|
Bulbtronics™
2627 |
5.4 A | SFA9-2, SFA7.5-2 | 0.39 in. | 3.54 in. |
Each for $947.32
|
|
||
Specifications
Xenon Arc Lamp | |
Xenon | |
Xenon Arc |