Poltex™ Bulk Bin for Benchtop Stack System DFS Item
Part of the Benchtop Storage System, this benchtop storage bin with lid boasts inside dividers that are moveable and removable to provide options for what can be stored.
Supplier:  Poltex™ BTSYSBIN
Catalog No. 22-000-806
Explore available promotions
Description
Description
  • All products in the Benchtop Storage line can stack on one another using our foot and pocket design to create ideal benchtop organization for your lab
  • Super durable and cleanable PETG construction
  • Rubber bumpons on base for non-slip placement
Specifications
Specifications
Lab Box Organizer
Clear
PETG