missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wanzl Bulk Shelving CF Back Label Holder

CF Back Label Holder for bulk shelving.

$12.00 - $14.77

Specifications

For Use With (Application) Shelving
Includes Label Holder
Color Black
Material Steel
View More Specs

Products 2
Catalog Number Mfr. No. Width (English) Price Quantity  
Catalog Number Mfr. No. Width (English) Price Quantity  
11222146
View Documents
Wanzl
TBCRLHB36B
36 in.
Each for $12.00
 
11222147
View Documents
Wanzl
TBCRLHB48B
48 in.
Each for $14.77
 
Description

Description

  • CF Back Label Holder
  • Color: Black
  • Accessory for bulk shelving system
Specifications

Specifications

Shelving
Label Holder
Black
Steel
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'