missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Wanzl Bulk Shelving Magnetic Label Holder
Magnetic Label Holder for use with bulk shelving.
$12.66 - $15.36
Specifications
For Use With (Application) | Shelving |
---|---|
Includes | Magnetic Label Holder |
Color | Black |
Material | Steel |
Description
- Magnetic Label Holder
- Color: Black
- Accessory for bulk shelving system
Specifications
Shelving | |
Magnetic Label Holder | |
Black | |
Steel |