missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Air-Tite™ Bulk Unsterile Syringes
Air-Tite™ Bulk Unsterile Syringes are high quality, yet economical for laboratory and veterinary use only
Supplier: Air-Tite™ MSB5
Description
- Bulk Disposable Syringes
- Smooth plunger
- Bold graduations
- Unsterile
- Polypropylene
- Economical
- Single-use
- Latex Free
- ISO
Specifications
Not Autoclavable | |
Polyisoprene Rubber (Tip), Polypropylene, Polypropylene (Barrel), Polypropylene (Plunger) | |
Laboratory/Veterinary | |
Luer Slip | |
Bulk | |
Leur Slip | |
Non-sterile |
ISO 13485 | |
Clear | |
Yes | |
Single-use | |
5 mL | |
Yes |
Not for human use - laboratory and veterinary use only