Air-Tite™ Bulk Unsterile Syringes
Air-Tite™ Bulk Unsterile Syringes are high quality, yet economical for laboratory and veterinary use only
Supplier:  Air-Tite™ MLB60
 View more versions of this product
Catalog No. 14-817-177
Explore available promotions
Description
Description
  • Bulk Disposable Syringes
  • Smooth plunger
  • Bold graduations
  • Unsterile
  • Polypropylene
  • Economical
  • Single-use
  • Latex Free
  • ISO
Specifications
Specifications
Not Autoclavable
Polyisoprene Rubber (Tip), Polypropylene, Polypropylene (Barrel), Polypropylene (Plunger)
Laboratory / Veterinary
Luer Lock
Bulk
Luer Lock
Non-sterile
ISO 13485
Clear
Yes
Single-use
60 mL
Yes
SDS
Documents
Documents
Promotions
Promotions
Not for human use - laboratory and veterinary use only