Bullard™ Breathing Air Supply Hose Nipple
Quick-disconnect nipples are for use with V10 airline hoses.
Supplier: Bullard™ 3422
Description
- Accessory for use with V10 hoses only
- 0.25 in. nipple
- Available in bayonet or schrader steel
Specifications
0.25 in. Bayonet | |
V10 hoses | |
3422 |
Hose Nipple | |
V12 hose adapter |